Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis)
Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis)
Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis)
Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis)
Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis)
Thermo Scientific Chemicals

Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis)

CAS: 15283-45-1 | AuNa3O6S4 | 490.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 039741.14
also known as 039741-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS15283-45-1
IUPAC Name{[(auriooxy)sulfonyl]sulfanyl}sodium; {[(sodiooxy)sulfonyl]sulfanyl}sodium
Molecular FormulaAuNa3O6S4
InChI KeyFJCMCHNDCIYVST-UHFFFAOYSA-J
SMILES[Na]OS(=O)(=O)S[Na].[Na]SS(=O)(=O)O[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (special)Au: ≥34.8%
FormPowder
Total Metal Impurities≤0.1%
Gold(I) sodium thiosulfate hydrate is useful in organogold compounds. Further, it serves as an antirheumatic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only