Silicon carbide, 400 grinding compound, 2oz (57g)
Silicon carbide, 400 grinding compound, 2oz (57g)
Silicon carbide, 400 grinding compound, 2oz (57g)
Thermo Scientific Chemicals

Silicon carbide, 400 grinding compound, 2oz (57g)

CAS: 409-21-2 | CSi | 40.10 g/mol
Have Questions?
Catalog NumberQuantity
039800.LA
also known as 039800-LA
57 g
Catalog number 039800.LA
also known as 039800-LA
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
83.70
Save 12.05 (14%)
Each
Quantity:
57 g
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
83.70
Save 12.05 (14%)
Each
Chemical Identifiers
CAS409-21-2
IUPAC Namemethanidylidynesilylium
Molecular FormulaCSi
InChI KeyHBMJWWWQQXIZIP-UHFFFAOYSA-N
SMILES[C-]#[Si+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black to gray
FormPowder
Particle Size400 Grit
Grinding compound for polishing IR crystal windows

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only