Cerium(IV) oxide, polishing compound, 57g (2oz)
Cerium(IV) oxide, polishing compound, 57g (2oz)
Cerium(IV) oxide, polishing compound, 57g (2oz)
Thermo Scientific Chemicals

Cerium(IV) oxide, polishing compound, 57g (2oz)

CAS: 1306-38-3 | CeO2 | 172.114 g/mol
Have Questions?
Catalog NumberQuantity
039802.LA
also known as 039802-LA
57 g
Catalog number 039802.LA
also known as 039802-LA
Price (USD)
86.65
Special offer
Online exclusive
Ends: 30-Jun-2026
102.00
Save 15.35 (15%)
Each
Quantity:
57 g
Request bulk or custom format
Price (USD)
86.65
Special offer
Online exclusive
Ends: 30-Jun-2026
102.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS1306-38-3
IUPAC Namedioxocerium
Molecular FormulaCeO2
InChI KeyCETPSERCERDGAM-UHFFFAOYSA-N
SMILESO=[Ce]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
FormPowder
Particle Sizeca. 3 micron
Polishing compound for IR crystal windows

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only