Palladium, 5% on strontium carbonate powder, reduced
Palladium, 5% on strontium carbonate powder, reduced
Palladium, 5% on strontium carbonate powder, reduced
Thermo Scientific Chemicals

Palladium, 5% on strontium carbonate powder, reduced

CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 039819.04
also known as 039819-04
Price (USD)
36.70
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
36.70
Each
Specifications
CAS5-3-7440
ChEBICHEBI:33363
Chemical Name or MaterialPalladium
IUPAC Namepalladium
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only