Analog 50 R ¾ in. Conductivity Meter
Analog 50 R ¾ in. Conductivity Meter
Actual product may vary
Thermo Scientific™
Analog 50 R ¾ in. Conductivity Meter
Record measurements for high purity water simply and reliably with Thermo Scientific™ Analog 50 R 3/4 in. Conductivity Meter.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
04.1612Stainless Steel Cartridge DI 750
04.1601Stainless Steel Cartridges 02.1500 to 02.15000
04.1608Stainless Steel Cartridge DI 425
Catalog number 04.1612
Price (USD)
-
Request A Quote
For Use With (Equipment):
Stainless Steel Cartridge DI 750
Record measurements for high purity water simply and reliably with Thermo Scientific™ Analog 50 R 3/4 in. Conductivity Meter.
  • Analog conductivity meter for a continuous control of quality
  • Measurement range of conductivity is 0 - 50μS/cm
  • Meter ships complete with 2 x 1.5m hoses, R 3/4 in. female thread and measure cell
Specifications
Inlet Connection NPTF (English)R 3/4 in.
Measurement Ranges0 to 50µS/cm
For Use With (Equipment)Stainless Steel Cartridge DI 750
TypeConductivity Meter, Analog
Unit SizeEach