Silver chromate, 99% min
Silver chromate, 99% min
Silver chromate, 99% min
Thermo Scientific Chemicals

Silver chromate, 99% min

CAS: 7784-01-2 | Ag2CrO4 | 331.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 040107.14
also known as 040107-14
Price (USD)
250.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
250.00
Each
Chemical Identifiers
CAS7784-01-2
IUPAC Namedisilver(1+) dioxochromiumbis(olate)
Molecular FormulaAg2CrO4
InChI KeyOJKANDGLELGDHV-UHFFFAOYSA-N
SMILES[Ag+].[Ag+].[O-][Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only