Phosphotungstic acid hydrate
Thermo Scientific Chemicals

Phosphotungstic acid hydrate

CAS: 12067-99-1 | P2O5·24WO3·xH2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 040116.36
also known as 040116-36
Price (USD)
582.65
Special offer
Online exclusive
Ends: 30-Jun-2026
685.00
Save 102.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
582.65
Special offer
Online exclusive
Ends: 30-Jun-2026
685.00
Save 102.35 (15%)
Each
Chemical Identifiers
CAS12067-99-1
SpecificationsSpecification SheetSpecification Sheet
Impurity content0.5% max
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphotungstic acid is used as a catalyst for the hydrolysis of propene to prepare 2-propanol. It is also involved in heterogeneous catalysis reaction to prepare propene from 2-propanol and methanol from hydrocarbons. Its ethanolic solution is used in biology as well as to precipitate proteins. It is utilized to precipitate different types of dye such as basic dyes and triphenylmethane dyes. In histology, it is used for staining of cell specimens. It is used in the gravimetric analysis of potassium in analytical chemistry.

Solubility
Soluble in water, alcohol and ether.

Notes
Incompatible with strong bases, strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhao, S.; Xu, G.; Chang, J.; Chang, C.; Bai, J.; Fang, S.; Liu, Z. Direct Production of Ethyl Levulinate from Carbohydrates Catalyzed by H-ZSM-5 Supported Phosphotungstic Acid. Bioresources 2015, 10 (2), 2223-2234.
  2. Villa, K.; Murcia-López, S.; Andreu, T.; Morante, J. R. Mesoporous WO3 photocatalyst for the partial oxidation of methane to methanol using electron scavengers. Appl. Catal., B 2015, 163, 150-155.