Copper(II) trifluoromethanesulfonate, 99%
Copper(II) trifluoromethanesulfonate, 99%
Copper(II) trifluoromethanesulfonate, 99%
Thermo Scientific Chemicals

Copper(II) trifluoromethanesulfonate, 99%

CAS: 34946-82-2 | CHCuF3O3S | 213.617 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
100 g
Catalog number 040133.18
also known as 040133-18
Price (USD)
441.65
Online Exclusive
491.00
Save 49.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
441.65
Online Exclusive
491.00
Save 49.35 (10%)
Each
Chemical Identifiers
CAS34946-82-2
IUPAC Namecopper(II) triflate
Molecular FormulaC2H2CuF6O4S2
InChI KeyTXWAGEFXPDWAQH-UHFFFAOYSA-N
SMILES[Cu].FC(F)(F)S(=O)=O.FC(F)(F)S(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)≥98.5%
FormPowder
Appearance (Color)White
Copper(II) trifluoromethanesulfonate is a mild lewis acid. It is used as catalyst which promotes dehydration of alcohols and diols to alkenes at ambient temperatures. It is widely used to generate carbenoid species from ?-diazo esters and ketones, via in situ reduction to the Cu(I) species. It is also promotes the reaction between diazo esters and imines to give aziridines. It catalyzes syn-selective aldol condensation of (Z)-silyl enol ethers with aldehydes, Friedel-Crafts alkylation, acylation reactions of aromatics and addition of trimethylsilyl cyanide to carbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only