Calcium trifluoromethanesulfonate, 99% min
Calcium trifluoromethanesulfonate, 99% min
Calcium trifluoromethanesulfonate, 99% min
Thermo Scientific Chemicals

Calcium trifluoromethanesulfonate, 99% min

CAS: 55120-75-7 | C2CaF6O6S2 | 338.204 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
100 g
Catalog number 040134.09
also known as 040134-09
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
106.00
Save 15.35 (14%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
106.00
Save 15.35 (14%)
Each
Chemical Identifiers
CAS55120-75-7
IUPAC Namecalcium ditrifluoromethanesulfonate
Molecular FormulaC2CaF6O6S2
InChI KeyPUQLFUHLKNBKQQ-UHFFFAOYSA-L
SMILES[Ca++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99% min
Calcium trifluoromethanesulfonate is used for aminolysis of epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only