Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min
Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min
Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min
Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min
Thermo Scientific Chemicals

Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min

CAS: 10378-47-9 | (NH4)4Ce(SO4)4·2H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 040194.30
also known as 040194-30
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS10378-47-9
IUPAC Nameλ⁴-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
Molecular FormulaCeH20N4O18S4
InChI KeyVCNAMBGKEDPVGQ-UHFFFAOYSA-J
SMILESN.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to yellow
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥94.0% (NH₄)₄Ce(SO₄)₄•2H₂O
Impurity contentInsoluble in dilute sulfuric acid: ≤0.05%
Impurity contentIron (Fe): ≤0.01%
View more
Ammonium cerium(IV) sulfate dihydrate is an oxidizing agent in organic synthesis and it is used as an analytical reagent, in thin-layer chromatography for the detection of anti-depressant drugs in human blood and urine samples. It is used to wet etch the Co catalyst and synthesis of carbon Nanotubes. It is used in medical and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only