Cobalt(II) tungsten oxide, 99%
Cobalt(II) tungsten oxide, 99%
Cobalt(II) tungsten oxide, 99%
Thermo Scientific Chemicals

Cobalt(II) tungsten oxide, 99%

pigment, drier for enamels, inks, and paints, electronic devices | CAS: 10101-58-3 | CoO4W | 306.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 040223.09
also known as 040223-09
Price (USD)
45.65
Special offer
Online exclusive
Ends: 30-Jun-2026
53.60
Save 7.95 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
45.65
Special offer
Online exclusive
Ends: 30-Jun-2026
53.60
Save 7.95 (15%)
Each
Chemical Identifiers
CAS10101-58-3
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
It is used as a pigment, drier for enamels, inks, and paints, electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only