Cobalt(II) selenide, 99+% (metals basis), Thermo Scientific Chemicals
Cobalt(II) selenide, 99+% (metals basis), Thermo Scientific Chemicals
Cobalt(II) selenide, 99+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) selenide, 99+% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
040224.18
also known as 040224-18
50 g
Catalog number 040224.18
also known as 040224-18
Price (USD)
213.65
Online Exclusive
237.00
Save 23.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
213.65
Online Exclusive
237.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS1307-99-9
IUPAC Nameselanylidenecobalt
Molecular FormulaCoSe
InChI KeyQVYIMIJFGKEJDW-UHFFFAOYSA-N
SMILES[Co]=[Se]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only