Calcium hydrogen phosphate dihydrate, 98% min
Calcium hydrogen phosphate dihydrate, 98% min
Calcium hydrogen phosphate dihydrate, 98% min
Calcium hydrogen phosphate dihydrate, 98% min
Thermo Scientific Chemicals

Calcium hydrogen phosphate dihydrate, 98% min

CAS: 7789-77-7 | CaH5O6P | 172.086 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 040233.A7
also known as 040233-A7
Price (USD)
245.65
Special offer
Online exclusive
Ends: 30-Jun-2026
289.00
Save 43.35 (15%)
Each
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)
245.65
Special offer
Online exclusive
Ends: 30-Jun-2026
289.00
Save 43.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7789-77-7
IUPAC Namecalcium dihydrate hydrogen phosphate
Molecular FormulaCaH5O6P
InChI KeyXAAHAAMILDNBPS-UHFFFAOYSA-L
SMILESO.O.[Ca++].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98% min
Calcium hydrogen phosphate dihydrate is used in the manufacture of pharmaceutical tablets and as a source of calcium and phosphorus in nutritional supplements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only