Thermo Scientific Chemicals

Strontium molybdenum oxide, 99%, Thermo Scientific Chemicals

Catalog number: 040238.09
10 g, Each
Thermo Scientific Chemicals

Strontium molybdenum oxide, 99%, Thermo Scientific Chemicals

Catalog number: 040238.09
10 g, Each
Quantity
Catalog number: 040238.09
also known as 040238-09
Price (USD)
Price: 23.50
Online price: 20.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13470-04-7
Molecular Formula
MoO4Sr
InChI Key
YZJCJRRZBDFNQA-UHFFFAOYSA-N
SMILES
[Sr++].[O-][Mo]([O-])(=O)=O
Molecular Weight (g/mol)
247.57
Form
Powder
Assay (unspecified)
>98.5%

Description

It is used as an optical material for a small range of special applications, for example, as an optical coating on lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text