Selenium(I) chloride, 99%
Selenium(I) chloride, 99%
Selenium(I) chloride, 99%
Thermo Scientific Chemicals

Selenium(I) chloride, 99%

CAS: 10025-68-0 | Cl2Se2 | 228.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
2 x 25 g
Catalog number 040239.14
also known as 040239-14
Price (USD)
414.65
Online Exclusive
460.00
Save 45.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
414.65
Online Exclusive
460.00
Save 45.35 (10%)
Each
Chemical Identifiers
CAS10025-68-0
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Selenium monochloride is used in electronics, instrumentation and other industrial purposes. It is used as a reagent for the synthesis of Se-containing compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only