Lithium manganese(III,IV) oxide, 99.5% (metals basis)
Lithium manganese(III,IV) oxide, 99.5% (metals basis)
Lithium manganese(III,IV) oxide, 99.5% (metals basis)
Lithium manganese(III,IV) oxide, 99.5% (metals basis)
Thermo Scientific Chemicals

Lithium manganese(III,IV) oxide, 99.5% (metals basis)

CAS: 12057-17-9 | LiMn2O4 | 180.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 040250.14
also known as 040250-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS12057-17-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue to black
FormPowder, -325 mesh
CommentStabilizer: Aluminum
Total Metal Impurities≤0.5%
It is used as anode materials of high-performance lithium ion battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only