Ammonium iodide, 99.995% (metals basis)
Ammonium iodide, 99.995% (metals basis)
Ammonium iodide, 99.995% (metals basis)
Thermo Scientific Chemicals

Ammonium iodide, 99.995% (metals basis)

Light Sensitive and Hygroscopic | CAS: 12027-06-4 | H4IN | 144.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 040255.22
also known as 040255-22
Price (USD)
918.65
Online Exclusive
1,020.00
Save 101.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
918.65
Online Exclusive
1,020.00
Save 101.35 (10%)
Each
Chemical Identifiers
CAS12027-06-4
IUPAC Nameammonium iodide
Molecular FormulaH4IN
InChI KeyUKFWSNCTAHXBQN-UHFFFAOYSA-N
SMILES[NH4+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (special)Iodine: 85.5-89.5%
Total Metal Impurities≤0.005% (50 ppm)
FormCrystals
It is used in photographic chemicals and some medications. Ammonium iodide and hydrogen peroxide plays an essential role for the eco-friendly oxyiodination of aromatic compounds. Ammonium Iodide and DMF is also useful for the copper-mediated selective cyanation reaction of Indoles and 2-Phenylpyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only