Dysprosium rod, 6.35mm (0.25in) dia, 99.7% (metals basis excluding Ta)
Dysprosium rod, 6.35mm (0.25in) dia, 99.7% (metals basis excluding Ta)
Dysprosium rod, 6.35mm (0.25in) dia, 99.7% (metals basis excluding Ta)
Thermo Scientific Chemicals

Dysprosium rod, 6.35mm (0.25in) dia, 99.7% (metals basis excluding Ta)

Form: Approx. 0.27g/mm, Cast rod, diameter is approximate | CAS: 7429-91-6 | Dy | 162.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
25 mm
50 mm
100 mm
Catalog number 040285.DJ
also known as 040285-DJ
Price (USD)
357.65
Special offer
Online exclusive
Ends: 30-Jun-2026
420.00
Save 62.35 (15%)
Each
Quantity:
50 mm
Request bulk or custom format
Price (USD)
357.65
Special offer
Online exclusive
Ends: 30-Jun-2026
420.00
Save 62.35 (15%)
Each
Chemical Identifiers
CAS7429-91-6
IUPAC Namedysprosium
Molecular FormulaDy
InChI KeyKBQHZAAAGSGFKK-UHFFFAOYSA-N
SMILES[Dy]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Ta0.3% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only