Gadolinium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)
Gadolinium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)
Gadolinium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)
Thermo Scientific Chemicals

Gadolinium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)

CAS: 7440-54-2 | Gd | 157.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
25 mm
Catalog number 040290.DH
also known as 040290-DH
Price (USD)
676.65
Online Exclusive
752.00
Save 75.35 (10%)
Each
Quantity:
25 mm
Request bulk or custom format
Price (USD)
676.65
Online Exclusive
752.00
Save 75.35 (10%)
Each
Chemical Identifiers
CAS7440-54-2
IUPAC Namegadolinium
Molecular FormulaGd
InChI KeyUIWYJDYFSGRHKR-UHFFFAOYSA-N
SMILES[Gd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Ta0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only