Neodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta)
Neodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta)
Neodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta)
Thermo Scientific Chemicals

Neodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta)

CAS: 7440-00-8 | Nd | 144.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
25 mm
50 mm
100 mm
Catalog number 040294.DH
also known as 040294-DH
Price (USD)
199.65
Special offer
Online exclusive
Ends: 28-Dec-2026
235.00
Save 35.35
Each
Quantity:
25 mm
Request bulk or custom format
Price (USD)
199.65
Special offer
Online exclusive
Ends: 28-Dec-2026
235.00
Save 35.35
Each
Chemical Identifiers
CAS7440-00-8
IUPAC Nameneodymium
Molecular FormulaNd
InChI KeyQEFYFXOXNSNQGX-UHFFFAOYSA-N
SMILES[Nd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Ta0.5% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only