Terbium(III) trifluoromethanesulfonate, 98%
Terbium(III) trifluoromethanesulfonate, 98%
Terbium(III) trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals

Terbium(III) trifluoromethanesulfonate, 98%

CAS: 148980-31-8 | C3F9O9S3Tb | 606.115 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 040312.04
also known as 040312-04
Price (USD)
88.30
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
88.30
Each
Chemical Identifiers
CAS148980-31-8
IUPAC Nameterbium(3+) tritrifluoromethanesulfonate
Molecular FormulaC3F9O9S3Tb
InChI KeyJLYAVYSVLLIWEK-UHFFFAOYSA-K
SMILES[Tb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder

It is used as a reagent & chemical intermediates. It is a raw material used in synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only