Yttrium(III) trifluoromethanesulfonate hydrate
Yttrium(III) trifluoromethanesulfonate hydrate
Yttrium(III) trifluoromethanesulfonate hydrate
Thermo Scientific Chemicals

Yttrium(III) trifluoromethanesulfonate hydrate

CAS: 34629-25-9 | C3F9O9S3Y | 536.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 040315.04
also known as 040315-04
Price (USD)
34.00
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
34.00
Each
Chemical Identifiers
CAS34629-25-9
IUPAC Nameyttrium(3+) tritrifluoromethanesulfonate
Molecular FormulaC3F9O9S3Y
InChI KeyJPJIEXKLJOWQQK-UHFFFAOYSA-K
SMILES[Y+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Y 12% min

It is employed in preparation of superconducting thin films. It is used in chemical research & as a intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only