Potassium heptafluorotantalate(V), 99.7%
Potassium heptafluorotantalate(V), 99.7%
Potassium heptafluorotantalate(V), 99.7%
Thermo Scientific Chemicals

Potassium heptafluorotantalate(V), 99.7%

CAS: 16924-00-8 | F7K2Ta | 392.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 040316.18
also known as 040316-18
Price (USD)
184.65
Online exclusive
205.00
Save 20.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
184.65
Online exclusive
205.00
Save 20.35
Each
Chemical Identifiers
CAS16924-00-8
IUPAC Namedipotassium heptafluorotantalumdiuide
Molecular FormulaF7H7K2Ta
InChI KeyBLXHHPZXEPALDX-UHFFFAOYSA-N
SMILESF.F.F.F.F.F.F.[K+].[K+].[Ta--]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.7% min
It is an important raw material. Used in chemical research. Use as primary and secondary intermediate(Pharmaceutical Intermediates). Used in the production and preparation of tantalum powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only