Indium spheres, 1mm (0.04in) dia, 99.99% (metals basis)
Indium spheres, 1mm (0.04in) dia, 99.99% (metals basis)
Indium spheres, 1mm (0.04in) dia, 99.99% (metals basis)
Thermo Scientific Chemicals

Indium spheres, 1mm (0.04in) dia, 99.99% (metals basis)

Tolerance: ±0.1mm | CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
Catalog number 040339.06
also known as 040339-06
Price (USD)
88.80
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
88.80
Each
Specifications
AppearanceSilvery-white spheres
Assay Percent Range99.99% (Metals basis)
CAS7440-74-6
ChEBICHEBI:30430
Chemical Name or MaterialIndium
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only