Sulfur in Heavy Mineral Oil standard solution, Specpure™, 30,000μg/g (3.00%)
Thermo Scientific Chemicals

Sulfur in Heavy Mineral Oil standard solution, Specpure™, 30,000μg/g (3.00%)

 
Have Questions?
Catalog NumberQuantity
040359.AE
also known as 040359-AE
100 mL
Catalog number 040359.AE
also known as 040359-AE
Price (USD)
108.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
108.00
Each
Specifications
Chemical Name or MaterialSulfur in Heavy Mineral Oil standard solution
Health Hazard 1H304-H315-H319-H335
Health Hazard 2GHS H Statement
H350-H315-H319
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280-P301+P310-P302+P352-P305+P351+P338-P331-P332+P313-P362-P501c
MDL NumberMFCD00804270
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only