Antimony(III) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Antimony(III) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Antimony(III) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimony(III) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
040372.04
also known as 040372-04
2 g
Catalog number 040372.04
also known as 040372-04
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.70
Save 14.05 (15%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.70
Save 14.05 (15%)
Each
Chemical Identifiers
CAS10025-91-9
IUPAC Nameantimony(3+) trichloride
Molecular FormulaCl3Sb
InChI KeyFAPDDOBMIUGHIN-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Sb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Antimony(III) chloride is used as a chlorinating agent, as a fireproofing agent in textiles; in bronzing steel and as a mordant in dyeing as well as a caustic in medicine. It is also used as an apoptosis agent that is used to detect cholesterols and carotenoids. It is also employed to detect certain compounds, such as cholesterols and carotenoids. This agent seems to cause dehydration of cholesterol to 3,5-cholestadiene and 2,4-cholestadiene which produce the blue color seen during the reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only