Silica gel desiccant, indicating, ACS - FAQs
Silica gel desiccant, indicating, ACS - FAQs
View additional product information for Silica gel desiccant, indicating, ACS - FAQs (040381.36, 040381.A4, 040381.22)
2 product FAQs found
Can I regenerate Silica gel desiccant, indicating, ACS?
Yes, this silica gel desiccant can be regenerated. This is achieved by heating in an oven at ~130 degrees C for 3 hours, or until the product returns to a blue color.
What color change indicates the Silica gel desiccant, indicating, ACS has reached moisture capacity?
The silica gel desiccant blue indicator spheres turn pink when they reach moisture capacity.