Bismuth citrate, 94%
Bismuth citrate, 94%
Bismuth citrate, 94%
Bismuth citrate, 94%
Thermo Scientific Chemicals

Bismuth citrate, 94%

CAS: 813-93-4 | C6H5BiO7 | 398.079 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 040398.30
also known as 040398-30
Price (USD)
113.00
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
113.00
Each
Add to cart
Chemical Identifiers
CAS813-93-4
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤1.0%
Assay from Suppliers CofAArsenic: Report value
Assay from Suppliers CofACopper: Report value
Assay from Suppliers CofALead: Report value
Assay from Suppliers CofANitrate: Report value
View more
Bismuth citrate is used as a progressive hair dye in cosmetics. It is used as antimicrobial agents for syphills, colitis, wound infection, quartan malaria, dyspepsia and peptic ulcers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only