Gold(I) chloride, Premion™, 99.99% (metals basis), Au 84.2% min
Gold(I) chloride, Premion™, 99.99% (metals basis), Au 84.2% min
Gold(I) chloride, Premion™, 99.99% (metals basis), Au 84.2% min
Thermo Scientific Chemicals

Gold(I) chloride, Premion™, 99.99% (metals basis), Au 84.2% min

CAS: 10294-29-8 | AuCl | 232.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number 040432.01
also known as 040432-01
Price (USD)
-
Quantity:
0.25 g
Request bulk or custom format
Chemical Identifiers
CAS10294-29-8
IUPAC Namegold(1+) chloride
Molecular FormulaAuCl
InChI KeyFDWREHZXQUYJFJ-UHFFFAOYSA-M
SMILES[Cl-].[Au+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Au 84.2% min
Total Metal Impurities0.01% max.
As intermediates. Also In Chemicals, pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only