Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), 98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), 98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), 98%
Thermo Scientific™

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), 98%

NMR shift reagent, nickel oxide precursor | CAS: 14481-08-4 | C22H38NiO4 | 425.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 040470.06
also known as 040470-06
Price (USD)
476.65
Online Exclusive
529.00
Save 52.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
476.65
Online Exclusive
529.00
Save 52.35 (10%)
Each
Specifications
CAS14481-08-4
Chemical Name or MaterialBis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II)
Formula Weight425.24
Health Hazard 1H317-H350
Health Hazard 2GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
View more
NMR shift reagent, nickel oxide precursor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only