Erbium(III) acetate tetrahydrate, REacton™, 99.9% (REO)
Erbium(III) acetate tetrahydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Erbium(III) acetate tetrahydrate, REacton™, 99.9% (REO)

CAS: 15280-57-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 040481.06
also known as 040481-06
Price (USD)
44.65
Special offer
Online exclusive
Ends: 31-Dec-2025
52.80
Save 8.15 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 31-Dec-2025
52.80
Save 8.15 (15%)
Each
Add to cart
Chemical Identifiers
CAS15280-57-6
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Erbium(III) acetate tetrahydrate participates in the synthesis and determination of crystal structures and stereochemical properties of lanthanide(III) complexes with ethylenediamine-N,N,N?,N?-tetraacetate. Participates in simple one-pot single-step synthesis of rare earth doped spherical mesoporous metal oxide nanospheres with upconversion emission ability in supercritical methanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only