Iron rod, 12mm (0.47in) dia, 99.99% (metals basis)
Iron rod, 12mm (0.47in) dia, 99.99% (metals basis)
Iron rod, 12mm (0.47in) dia, 99.99% (metals basis)
Thermo Scientific Chemicals

Iron rod, 12mm (0.47in) dia, 99.99% (metals basis)

CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
50 mm
Catalog number 040499.DJ
also known as 040499-DJ
Price (USD)
521.65
Online Exclusive
579.00
Save 57.35 (10%)
Each
Quantity:
50 mm
Request bulk or custom format
Price (USD)
521.65
Online Exclusive
579.00
Save 57.35 (10%)
Each
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod, 12mm (0.47in) diameter.
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.99% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only