Tungsten carbide, 99% (metals basis)
Tungsten carbide, 99% (metals basis)
Tungsten carbide, 99% (metals basis)
Thermo Scientific Chemicals

Tungsten carbide, 99% (metals basis)

CAS: 12070-12-1 | CW | 195.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 040502.18
also known as 040502-18
Price (USD)
56.65
Special offer
Online exclusive
Ends: 28-Dec-2026
66.70
Save 10.05
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
56.65
Special offer
Online exclusive
Ends: 28-Dec-2026
66.70
Save 10.05
Each
Chemical Identifiers
CAS12070-12-1
IUPAC Namemethanidylidynetungstenylium
Molecular FormulaCW
InChI KeyUONOETXJSWQNOL-UHFFFAOYSA-N
SMILES[C-]#[W+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Sintered tungsten carbide cutting tools are very abrasion resistant. Tungsten carbide is often used in armor-piercing ammunition. Tungsten carbide is also an effective neutron reflector. Hard carbides, especially tungsten carbide, are used by athletes for balance. Tungsten carbide is also used for making surgical instruments

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only