Samco™ Narrow Mouth Bio-Tite™ 40mL (1 oz.) 48mm Specimen Containers
Samco™ Narrow Mouth Bio-Tite™ 40mL (1 oz.) 48mm Specimen Containers
Samco™ Narrow Mouth Bio-Tite™ 40mL (1 oz.) 48mm Specimen Containers
Samco™ Narrow Mouth Bio-Tite™ 40mL (1 oz.) 48mm Specimen Containers
Thermo Scientific™

Samco™ Narrow Mouth Bio-Tite™ 40mL (1 oz.) 48mm Specimen Containers

Prevent bio-hazardous spills and conserve storage space with these stackable containers, featuring a proprietary, all plastic, leak-resistant seal.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoColor del tapón
040506
también denominado 04 0506
Amarillo
040181
también denominado 04 0181
Blanco
Número de catálogo 040506
también denominado 04 0506
Precio (MXN)
-
Color del tapón:
Amarillo
Prevent bio-hazardous spills and conserve storage space with stackable Thermo Scientific™ Samco™ Narrow-Mouth Bio-Tite™ 40mL (1 oz.) 48mm Specimen Containers. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.
Features:
  • Dual-threaded caps create leak-resistant seal for superior safety and handling
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Virgin polypropylene allows higher clarity to see samples, and will not become brittle
  • Sterile indicator tab assures tamper evidence
  • Bilingual English and Spanish label facilitates accurate usage

Ordering Information:

  • Available nonsterile or gamma irradiation sterilized
  • Available labled or non-labled
  • Available bulk or individually packed
  • In cases of 500
Especificaciones
Capacidad (métrico)40 mL
Color del tapónAmarillo
Tamaño del tapón48 mm
Tipo de tapónBio-Tite Lid
DescripciónCap and container bagged separately
Diámetro (métrico)48 mm
EtiquetaNon-tabbed, 10% buffered formalin
N.º por caja600/Cs.
N.° por paquete600/Pk.
Closure IncludedSí
MaterialVirgin Polypropylene
Tipo de productoSpecimen Container
FormaCilíndrico
EsterilidadNo estéril
Unit SizeCase of 600