Niobium powder, -325 mesh, Puratronic™, 99.99% (metals basis excluding Ta), Ta ∣<500ppm
Niobium powder, -325 mesh, Puratronic™, 99.99% (metals basis excluding Ta), Ta ∣<500ppm
Niobium powder, -325 mesh, Puratronic™, 99.99% (metals basis excluding Ta), Ta ∣<500ppm
Thermo Scientific Chemicals

Niobium powder, -325 mesh, Puratronic™, 99.99% (metals basis excluding Ta), Ta ∣<500ppm

CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040510.09
also known as 040510-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to dark grey
FormPowder or crystals
Total Metal Impurities excluding Ta≤0.01% max.
Particle Size-325 mesh ( less than 45 micron )
Trace MetalTotal metal impurities less than 0.01% excluding Ta, Ta <500ppm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only