Bismuth Lead Tin ingot (Rose's metal)
Bismuth Lead Tin ingot (Rose's metal)
Bismuth Lead Tin ingot (Rose's metal)
Thermo Scientific Chemicals

Bismuth Lead Tin ingot (Rose's metal)

CAS: 56001-37-7 | C10H14N5Na2O14P3 | 567.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040516.22
also known as 040516-22
Price (USD)
160.65
Online Exclusive
178.00
Save 17.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
160.65
Online Exclusive
178.00
Save 17.35 (10%)
Each
Specifications
Assay Percent Notes(Rose's metal)
Avg. Mol. Wt. or Mol. Wt. RangeBi:Pb:Sn; 50:25:25 wt%
CAS56001-37-7
Chemical Name or MaterialBismuth Lead Tin ingot (Rose's metal)
Health Hazard 1H302+H332-H360Df-H373
View more
Rose's metal was used as the droplet fluid, because it has a low melting point (e460 K). Rose's metal is used as a sample in computer-controlled diffractometer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only