Ethylenediaminetetraacetic acid manganese disodium salt hydrate
Ethylenediaminetetraacetic acid manganese disodium salt hydrate
Ethylenediaminetetraacetic acid manganese disodium salt hydrate
Ethylenediaminetetraacetic acid manganese disodium salt hydrate
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid manganese disodium salt hydrate

Metal chelating compound | CAS: 15375-84-5 | C10H12MnN2Na2O8 | 389.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 040519.14
also known as 040519-14
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS15375-84-5
IUPAC Namemanganese(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Molecular FormulaC10H12MnN2Na2O8
InChI KeyZQHKAIDDHTYINE-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Mn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>/= 95.0% ( anhyd. basis)
CommentPowder

Ethylenediaminetetraacetic acid manganese disodium salt hydrate is a metal chelating compound. Cobalt was analysed complexometrically using ethylenediaminetetra-acetic acid (EDTA) by masking aluminum. Previous studies of manganese-EDTA and manganese-EGT-1 complexes indicated that the &-nitrogen coordination of EDTA resulted in a highly anisotropic spectrum for Mn(II). Lipophilic derivatives of manganese EDTA chelates when incorporated into liposomes confer the greatest hepatic enhancement per micromole of metal ion and have favorable clearance kinetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only