Sulfur in Kerosene standard solution, Specpure™, 100μg/g (0.010%)
Thermo Scientific Chemicals

Sulfur in Kerosene standard solution, Specpure™, 100μg/g (0.010%)

 
Have Questions?
Catalog NumberQuantity
040537.AE
also known as 040537-AE
100 mL
Catalog number 040537.AE
also known as 040537-AE
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Specifications
Chemical Name or MaterialSulfur in Kerosene standard solution
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KEROSENE
Health Hazard 1H227-H304
Health Hazard 2GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.
Health Hazard 3P210-P280-P301+P310-P331-P370+P378q-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only