Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade
Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade
Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade
Thermo Scientific Chemicals

Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade

CAS: 14692-15-0 | Cl3ErH12O18 | 573.687 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
∼50 g
∼2 g
10 g
Catalog number 040576.NE
also known as 040576-NE
Price (USD)
629.65
Online Exclusive
699.00
Save 69.35 (10%)
Each
Quantity:
∼50 g
Request bulk or custom format
Price (USD)
629.65
Online Exclusive
699.00
Save 69.35 (10%)
Each
Chemical Identifiers
CAS14692-15-0
IUPAC Nameerbium(3+) hexahydrate triperchlorate
Molecular FormulaCl3ErH12O18
InChI KeyJZRNHNTYRRYOCR-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Er+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration (weight/weight)50% min
Erbium(III) perchlorate hexahydrate is used in nuclear technology for neutron-absorbing control rods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only