Yttrium(III) oxide pellets, 99.9% (REO), Thermo Scientific Chemicals
Yttrium(III) oxide pellets, 99.9% (REO), Thermo Scientific Chemicals
Yttrium(III) oxide pellets, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium(III) oxide pellets, 99.9% (REO), Thermo Scientific Chemicals

Sintered pellets suitable for vacuum deposition
Have Questions?
Catalog NumberQuantity
040648.22
also known as 040648-22
100 g
Catalog number 040648.22
also known as 040648-22
Price (USD)
420.65
Online Exclusive
467.00
Save 46.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
420.65
Online Exclusive
467.00
Save 46.35 (10%)
Each
Chemical Identifiers
CAS1314-36-9
IUPAC Namediyttrium(3+) trioxidandiide
Molecular FormulaO3Y2
InChI KeyRUDFQVOCFDJEEF-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Y+3].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Sintered pellets suitable for vacuum depositionYttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is a precursor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only