Lithium bromide, anhydrous, 99.995% (metals basis)
Lithium bromide, anhydrous, 99.995% (metals basis)
Lithium bromide, anhydrous, 99.995% (metals basis)
Thermo Scientific Chemicals

Lithium bromide, anhydrous, 99.995% (metals basis)

CAS: 7550-35-8 | BrLi | 86.844 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 040657.14
also known as 040657-14
Price (USD)
144.65
Online exclusive
161.00
Save 16.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
144.65
Online exclusive
161.00
Save 16.35
Each
Chemical Identifiers
CAS7550-35-8
IUPAC Namelithium(1+) bromide
Molecular FormulaBrLi
InChI KeyAMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES[Li+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
It is used for reconstitution of brines, as swelling agent for proteins and as electrolyte component in lithium battery. Used as brazing and welding flux. Lithium bromide is used as catalyst for the condensation of carbonyl compounds with active methylene compounds in the absence of solvent, operating medium for air-conditioning and industrial drying system. Also used in manufacturing pharmaceuticals and alkylation process. It also plays an important role in the synthesis of dihydropyrimidinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only