Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Thermo Scientific Chemicals

Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)

CAS: 64438-53-5 | C15H21O6Sm | 447.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 040665.06
also known as 040665-06
Price (USD)
40.10
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.10
Each
Chemical Identifiers
CAS64438-53-5
IUPAC Namesamarium(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21O6Sm
InChI KeyJTZJMDRBZZVNSP-UHFFFAOYSA-N
SMILES[Sm+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only