Nickel wire, 0.025mm (0.001in) dia, hard, 99.98% (metals basis)
Nickel wire, 0.025mm (0.001in) dia, hard, 99.98% (metals basis)
Nickel wire, 0.025mm (0.001in) dia, hard, 99.98% (metals basis)
Nickel wire, 0.025mm (0.001in) dia, hard, 99.98% (metals basis)
Thermo Scientific Chemicals

Nickel wire, 0.025mm (0.001in) dia, hard, 99.98% (metals basis)

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
50 cm
1 m
Catalog number 040672.BU
also known as 040672-BU
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.60
Save 7.95
Each
Quantity:
25 cm
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.60
Save 7.95
Each
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.025mm (0.001in) diameter, hardened.
Assay from Supplier's CofA≥99.98% (metals basis)
Appearance (Color)Silvery white
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only