Tantalum wire, 0.05mm (0.002in) dia, annealed, 99.9% (metals basis)
Tantalum wire, 0.05mm (0.002in) dia, annealed, 99.9% (metals basis)
Tantalum wire, 0.05mm (0.002in) dia, annealed, 99.9% (metals basis)
Thermo Scientific Chemicals

Tantalum wire, 0.05mm (0.002in) dia, annealed, 99.9% (metals basis)

Form: approx. 0.033g/m; Temper: soft | CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
10 m
Catalog number 040674.G6
also known as 040674-G6
Price (USD)
313.65
Online Exclusive
348.00
Save 34.35 (10%)
Each
Quantity:
10 m
Request bulk or custom format
Price (USD)
313.65
Online Exclusive
348.00
Save 34.35 (10%)
Each
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver Gray
FormWire, 0.05mm (0.002in) diameter, annealed.
Assay from Supplier's CofA≥99.9% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only