Tantalum wire, 0.025mm (0.001in) dia, hard, 99.9% (metals basis)
Tantalum wire, 0.025mm (0.001in) dia, hard, 99.9% (metals basis)
Tantalum wire, 0.025mm (0.001in) dia, hard, 99.9% (metals basis)
Thermo Scientific Chemicals

Tantalum wire, 0.025mm (0.001in) dia, hard, 99.9% (metals basis)

Form: Approx. 0.0082g/m; Temper: as drawn | CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
10 m
Catalog number 040675.G1
also known as 040675-G1
Price (USD)
179.65
Special offer
Online exclusive
Ends: 28-Dec-2026
211.00
Save 31.35
Each
Quantity:
1 m
Request bulk or custom format
Price (USD)
179.65
Special offer
Online exclusive
Ends: 28-Dec-2026
211.00
Save 31.35
Each
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray to Silver
FormWire, 0.025mm (0.001in) diameter, hardened.
Assay from Supplier's CofA≥99.9% (metals basis)
Microelectronics, contacts, surgical, high temperature

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only