Aluminum wire, 0.025mm (0.001in) dia, hard, 99.99% (metals basis)
Aluminum wire, 0.025mm (0.001in) dia, hard, 99.99% (metals basis)
Aluminum wire, 0.025mm (0.001in) dia, hard, 99.99% (metals basis)
Thermo Scientific Chemicals

Aluminum wire, 0.025mm (0.001in) dia, hard, 99.99% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Catalog NumberQuantity
040677.G11 m
Catalog number 040677.G1
Price (USD)
164.65
Special offer
Online exclusive
Ends: 30-Jun-2026
193.00
Save 28.35 (15%)
Each
Quantity:
1 m
Request bulk or custom format
Price (USD)
164.65
Special offer
Online exclusive
Ends: 30-Jun-2026
193.00
Save 28.35 (15%)
Each
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only