Aluminum Ultrathin foil, 0.8 micron thick, 99.1% (metals basis)
Aluminum Ultrathin foil, 0.8 micron thick, 99.1% (metals basis)
Aluminum Ultrathin foil, 0.8 micron thick, 99.1% (metals basis)
Aluminum Ultrathin foil, 0.8 micron thick, 99.1% (metals basis)
Thermo Scientific Chemicals

Aluminum Ultrathin foil, 0.8 micron thick, 99.1% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
30 x 30 mm
140 x 140 mm
Catalog number 040707.6X
also known as 040707-6X
Price (USD)
461.65
Online Exclusive
513.00
Save 51.35 (10%)
Each
Quantity:
140 x 140 mm
Request bulk or custom format
Price (USD)
461.65
Online Exclusive
513.00
Save 51.35 (10%)
Each
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormFoil, 0.8 micron thick.
Assay from Supplier's CofA≥99.1% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only