Palladium wire, 0.05mm (0.002 in.) dia., hard, 99.9% (metals basis), Thermo Scientific Chemicals
Palladium wire, 0.05mm (0.002 in.) dia., hard, 99.9% (metals basis), Thermo Scientific Chemicals
Palladium wire, 0.05mm (0.002 in.) dia., hard, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium wire, 0.05mm (0.002 in.) dia., hard, 99.9% (metals basis), Thermo Scientific Chemicals

Microelectronics, contacts
Have Questions?
Catalog NumberQuantity
040730.G1
also known as 040730-G1
1 m
Catalog number 040730.G1
also known as 040730-G1
Price (USD)
140.65
Special offer
Online exclusive
Ends: 28-Dec-2026
165.00
Save 24.35
Each
Quantity:
1 m
Request bulk or custom format
Price (USD)
140.65
Special offer
Online exclusive
Ends: 28-Dec-2026
165.00
Save 24.35
Each
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.9% (metals basis)
Appearance (Color)Silver Gray
FormWire, 0.05mm (0.002in) diameter, hardened.
Microelectronics, contacts

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only