Silver Thinfoil, 0.005mm (0.0002in) thick, 99.97% (metals basis)
Silver Thinfoil, 0.005mm (0.0002in) thick, 99.97% (metals basis)
Silver Thinfoil, 0.005mm (0.0002in) thick, 99.97% (metals basis)
Thermo Scientific Chemicals

Silver Thinfoil, 0.005mm (0.0002in) thick, 99.97% (metals basis)

Microelectronics, contacts and coatings | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 040748.FF
also known as 040748-FF
Price (USD)
377.00
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
377.00
Each
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.005mm (0.0002in) thick.
Assay from Supplier's CofA≥99.97% (metals basis)
Appearance (Color)Silver
Microelectronics, contacts and coatings

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only