Aluminum foil, 0.25mm (0.01in) thick, annealed, 99.99% (metals basis)
Aluminum foil, 0.25mm (0.01in) thick, annealed, 99.99% (metals basis)
Aluminum foil, 0.25mm (0.01in) thick, annealed, 99.99% (metals basis)
Thermo Scientific Chemicals

Aluminum foil, 0.25mm (0.01in) thick, annealed, 99.99% (metals basis)

Form: approx. 1.7g/50x50mm; Temper: soft | CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Catalog number 040761.FI
also known as 040761-FI
Price (USD)
35.65
Special offer
Online exclusive
Ends: 30-Jun-2026
41.90
Save 6.25 (15%)
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 30-Jun-2026
41.90
Save 6.25 (15%)
Each
Specifications
AppearanceSilver-colored foil
Assay Percent Range99.99% (Metals basis)
CAS7429-90-5
ChEBICHEBI:33629
Chemical Name or MaterialAluminum
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only